Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080888.1 | NP_543164.1 | 4 | 428 | ||
XM_006252091.2 | XP_006252153.1 | 4 | 2229 | |||
XM_008770733.1 | XP_008768955.1 | 4 | 2358 | |||
XM_008770734.1 | XP_008768956.1 | 4 | 2359 | |||
GenBank mRNA | AB077364.1 | - | 4 | 428 | ||
AF441118.1 | - | 4 | 497 | |||
BC081740.1 | - | 4 | 581 | |||
BC098658.1 | - | 4 | 462 | 7324593 | ||
FQ230363.1 | - | 4 | 421 | |||
FQ235252.1 | - | 4 | 447 |