Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133320.1 | NP_579854.1 | 3 | 221 | ||
XM_006246571.2 | XP_006246633.1 | 3 | 630 | |||
XM_006246572.2 | XP_006246634.1 | 3 | 623 | |||
XM_006246573.2 | XP_006246635.1 | 3 | 371 | |||
XM_006246574.2 | XP_006246636.1 | 3 | 256 | |||
XM_006246575.2 | XP_006246637.1 | 3 | 734 | |||
XM_006246576.2 | XP_006246638.1 | 3 | 624 | |||
XM_006246578.2 | XP_006246640.1 | 3 | 370 | |||
GenBank mRNA | AY008298.1 | - | 3 | 221 | ||
BC070909.1 | - | 3 | 311 | 7103650 |