Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALB SIRNA Assays ›
Gene Symbol:
Alb
Gene Name:
albumin
Gene Aliases:
Alb1, Albza
Chromosome Location:
Chr.14: 19176275 - 19191793 on Build Rnor_6.0
UniGene ID:
Rn.202968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS301987
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Alb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_134326.2
NP_599153.2
9
1128
GenBank mRNA
BC085359.1
-
9
1128
7303856
FQ209382.1
-
5
576
FQ209565.1
-
2
161
FQ209580.1
-
2
113
FQ209635.1
-
3
338
FQ209651.1
-
3
368
FQ209693.1
-
2
163
FQ209695.1
-
5
562
FQ209713.1
-
1
38
FQ209714.1
-
2
231
FQ209721.1
-
3
349
FQ209825.1
-
2
154
FQ209907.1
-
5
597
FQ209957.1
-
4
509
FQ209968.1
-
2
156
FQ210022.1
-
3
351
FQ210057.1
-
5
527
FQ210064.1
-
1
20
FQ210205.1
-
1
41
FQ210297.1
-
2
196
FQ210310.1
-
7
800
FQ210316.1
-
3
349
FQ210347.1
-
2
130
FQ210361.1
-
2
269
FQ210399.1
-
2
94
FQ210406.1
-
3
328
FQ210411.1
-
2
260
FQ210445.1
-
1
34
FQ210540.1
-
2
78
FQ210546.1
-
3
376
FQ210564.1
-
2
113
FQ210640.1
-
3
333
FQ210724.1
-
6
652
FQ210749.1
-
1
52
FQ210803.1
-
5
598
FQ210850.1
-
4
444
FQ210851.1
-
4
475
FQ210858.1
-
6
682
FQ210877.1
-
5
543
FQ210946.1
-
1
37
FQ210995.1
-
3
338
FQ211001.1
-
2
168
FQ211010.1
-
4
486
FQ211036.1
-
3
381
FQ211096.1
-
3
349
FQ211119.1
-
1
38
FQ211129.1
-
3
322
FQ211183.1
-
2
100
FQ211196.1
-
3
309
FQ211218.1
-
6
756
FQ211226.1
-
1
31
FQ211255.1
-
5
576
FQ211259.1
-
5
585
FQ211355.1
-
2
173
FQ211376.1
-
1
32
FQ211378.1
-
3
349
FQ211445.1
-
2
253
FQ218081.1
-
2
157
FQ218137.1
-
3
353
FQ218216.1
-
4
452
FQ218268.1
-
1
49
FQ218283.1
-
2
246
FQ218310.1
-
1
27
FQ218331.1
-
1
27
FQ218371.1
-
1
59
FQ218379.1
-
1
35
FQ218412.1
-
2
160
FQ218458.1
-
5
592
FQ218497.1
-
1
20
FQ218566.1
-
2
259
FQ218682.1
-
3
317
FQ218739.1
-
4
497
FQ218757.1
-
1
38
FQ218799.1
-
2
82
FQ218806.1
-
1
34
FQ218815.1
-
4
462
FQ218867.1
-
2
278
FQ218989.1
-
2
182
FQ219017.1
-
2
133
FQ219210.1
-
5
547
FQ219231.1
-
2
174
FQ219260.1
-
3
318
FQ219293.1
-
5
576
FQ219382.1
-
2
263
FQ219447.1
-
5
569
FQ219470.1
-
4
447
FQ219477.1
-
2
189
FQ219514.1
-
3
375
FQ219650.1
-
3
345
FQ219670.1
-
5
626
FQ219679.1
-
3
338
FQ219682.1
-
3
320
FQ219706.1
-
1
58
FQ219775.1
-
2
232
V01222.1
-
9
1113
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina homeostasis
transport
response to nutrient
cellular response to starvation
response to organic substance
hemolysis by symbiont of host erythrocytes
vasodilation
negative regulation of apoptotic process
positive regulation of circadian sleep/wake cycle, non-REM sleep
response to mercury ion
maintenance of mitochondrion location
response to platinum ion
DNA binding
fatty acid binding
drug binding
zinc ion binding
toxic substance binding
oxygen binding
enzyme binding
pyridoxal phosphate binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix