Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HPRT1 SIRNA Assays ›
          Gene Symbol:
          Hprt1 LOC103689983
          Gene Name:
          hypoxanthine phosphoribosyltransferase 1
          hypoxanthine-guanine phosphoribosyltransferase
          Gene Aliases:
          Hgprtase, Hprt
          -
          Chromosome Location:
          Chr.X: 158196640 - 158228815 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 158623231 - 158655274 on Build Rnor_6.0
          UniGene ID:
          Rn.47
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID RSS302238
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hprt1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012583.2 NP_036715.1 2, 3 194
          GenBank mRNA BC098629.1 - 2, 3 194 7386548
          M63983.1 - 2, 3 226
          S79292.1 - 2, 3 226
          X62085.1 - 154
          Gene Symbol: LOC103689983
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773659.1 XP_008771881.1 2, 3 270

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_013556, BC004686, AK002286, AK088114, AK082827, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          purine nucleotide biosynthetic process
          purine ribonucleoside salvage
          adenine salvage
          guanine salvage
          spermatogenesis
          brain development
          grooming behavior
          locomotory behavior
          cytolysis
          striatum development
          cerebral cortex neuron differentiation
          central nervous system neuron development
          GMP salvage
          IMP salvage
          cellular response to insulin stimulus
          dopamine metabolic process
          hypoxanthine salvage
          positive regulation of dopamine metabolic process
          GMP catabolic process
          IMP metabolic process
          adenine metabolic process
          hypoxanthine metabolic process
          lymphocyte proliferation
          dendrite morphogenesis
          protein homotetramerization
          nucleotide binding
          magnesium ion binding
          hypoxanthine phosphoribosyltransferase activity
          identical protein binding
          protein homodimerization activity
          guanine phosphoribosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b776c6558-ktt9x:80/100.66.134.75:80.
          git-commit: 08b7be5c3d3493be030128aff2ceec7cf49358c5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc