Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIPC SIRNA Assays ›
          Gene Symbol:
          Lipc
          Gene Name:
          lipase, hepatic
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 77272582 - 77398484 on Build Rnor_6.0
          UniGene ID:
          Rn.1195
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS302287
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lipc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012597.2 NP_036729.2 9 1557
          XM_006243359.2 XP_006243421.1 11 1660
          XM_006243360.2 XP_006243422.1 10 1612
          XM_006243361.2 XP_006243423.1 9 1429
          GenBank mRNA BC088160.1 - 9 1557 7323251
          M16235.1 - 9 1479
          X17366.1 - 9 1467

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          liver development
          fatty acid metabolic process
          fatty acid biosynthetic process
          triglyceride metabolic process
          phosphatidylserine metabolic process
          circadian rhythm
          cholesterol metabolic process
          response to hormone
          response to carbohydrate
          response to acetate
          response to inorganic substance
          response to organic cyclic compound
          heparan sulfate proteoglycan biosynthetic process
          triglyceride catabolic process
          cholesterol transport
          response to nutrient levels
          response to magnesium ion
          cellular response to hormone stimulus
          response to lipid
          chylomicron remodeling
          very-low-density lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          chylomicron remnant clearance
          low-density lipoprotein particle clearance
          response to drug
          cholesterol homeostasis
          response to amino acid
          response to peptide hormone
          phosphatidylethanolamine metabolic process
          neutral lipid catabolic process
          phosphatidylcholine metabolic process
          phosphatidic acid metabolic process
          glycerophospholipid catabolic process
          response to copper ion
          developmental growth
          protein oligomerization
          response to glucocorticoid
          response to calcium ion
          triglyceride homeostasis
          response to fatty acid
          regulation of plasma lipoprotein particle levels
          lysophospholipase activity
          triglyceride lipase activity
          heparin binding
          lipid binding
          phosphatidylcholine 1-acylhydrolase activity
          lipase activity
          transferase activity, transferring acyl groups
          low-density lipoprotein particle binding
          apolipoprotein binding
          chylomicron binding
          heparan sulfate proteoglycan binding
          acylglycerol lipase activity
          carboxylic ester hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0