Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP3CA SIRNA Assays ›
          Gene Symbol:
          Ppp3ca
          Gene Name:
          protein phosphatase 3, catalytic subunit, alpha isozyme
          Gene Aliases:
          Calna1
          Chromosome Location:
          Chr.2: 241909336 - 242186861 on Build Rnor_6.0
          UniGene ID:
          Rn.228648
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS302623
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ppp3ca
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017041.1 NP_058737.1 10 1304
          XM_006233363.2 XP_006233425.1 10 1799
          XM_006233364.2 XP_006233426.1 10 1064
          GenBank mRNA AY724517.1 - 10 1433
          D90035.1 - 10 1304
          FQ212355.1 - 10 1648
          FQ212817.1 - 5 355
          M29275.1 - 10 1304
          X57115.1 - 10 1367

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          response to amphetamine
          protein dephosphorylation
          protein import into nucleus
          calcium ion transport
          transition between fast and slow fiber
          cardiac muscle hypertrophy in response to stress
          dephosphorylation
          calcium-mediated signaling
          calcineurin-NFAT signaling cascade
          multicellular organismal response to stress
          negative regulation of chromatin binding
          cellular response to drug
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of calcium ion-dependent exocytosis
          negative regulation of insulin secretion
          skeletal muscle fiber development
          negative regulation of dendrite morphogenesis
          modulation of synaptic transmission
          positive regulation of NFAT protein import into nucleus
          response to calcium ion
          excitatory postsynaptic potential
          cellular response to glucose stimulus
          positive regulation of cardiac muscle hypertrophy in response to stress
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          calcium-dependent protein serine/threonine phosphatase activity
          protein binding
          calmodulin binding
          drug binding
          enzyme binding
          calmodulin-dependent protein phosphatase activity
          metal ion binding
          protein heterodimerization activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix