Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPARC SIRNA Assays ›
          Gene Symbol:
          Sparc
          Gene Name:
          secreted protein, acidic, cysteine-rich (osteonectin)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 40742390 - 40764222 on Build Rnor_6.0
          UniGene ID:
          Rn.234316
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS302733
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sparc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012656.1 NP_036788.1 8 519
          XM_008767663.1 XP_008765885.1 7 597
          GenBank mRNA BC061777.1 - 7 534 5621666
          D28875.1 - 7 536
          FQ212364.1 - 7 588
          FQ212499.1 - 7 553
          FQ213815.1 - 7 553
          FQ213945.1 - 7 553
          FQ214222.1 - 7 551
          FQ214492.1 - 7 553
          FQ214649.1 - 7 553
          FQ214673.1 - 7 553
          FQ215195.1 - 7 552
          FQ215581.1 - 7 554
          FQ215622.1 - 7 555
          FQ215818.1 - 7 564
          FQ215935.1 - 7 553
          FQ216823.1 - 7 555
          FQ217546.1 - 7 554
          FQ219866.1 - 7 535
          FQ219932.1 - 8 535
          FQ219983.1 - 7 553
          FQ220003.1 - 7 553
          FQ220043.1 - 7 551
          FQ220046.1 - 7 553
          FQ220186.1 - 7 551
          FQ220279.1 - 5 377
          FQ220287.1 - 7 555
          FQ220443.1 - 7 535
          FQ220507.1 - 7 561
          FQ220547.1 - 7 553
          FQ220572.1 - 7 553
          FQ220644.1 - 7 553
          FQ220827.1 - 7 553
          FQ220880.1 - 7 553
          FQ220901.1 - 7 553
          FQ222224.1 - 7 553
          FQ223025.1 - 7 553
          FQ224996.1 - 6 424
          FQ225033.1 - 7 555
          FQ225092.1 - 7 549
          FQ225200.1 - 7 553
          FQ225220.1 - 7 532
          FQ225486.1 - 7 535
          FQ225609.1 - 7 553
          FQ225618.1 - 7 551
          FQ225644.1 - 3 220
          FQ225678.1 - 7 553
          FQ225688.1 - 7 553
          FQ225694.1 - 7 555
          FQ225722.1 - 7 552
          FQ225884.1 - 7 551
          FQ225913.1 - 7 553
          FQ225914.1 - 7 555
          FQ225965.1 - 7 553
          FQ226299.1 - 7 535
          FQ226360.1 - 7 553
          FQ226584.1 - 7 553
          FQ226599.1 - 7 554
          FQ226651.1 - 7 576
          FQ226673.1 - 7 534
          FQ226746.1 - 7 551
          FQ227279.1 - 7 553
          FQ227628.1 - 7 562
          FQ227644.1 - 7 553
          FQ227708.1 - 7 553
          FQ227727.1 - 7 533
          FQ227749.1 - 7 553
          FQ228032.1 - 7 533
          FQ228597.1 - 7 535
          FQ229172.1 - 7 553
          FQ229199.1 - 7 553
          FQ229230.1 - 7 553
          FQ229283.1 - 7 535
          FQ229299.1 - 7 536
          FQ229315.1 - 7 551
          FQ229316.1 - 7 553
          FQ230029.1 - 7 553
          FQ230086.1 - 7 551
          FQ230210.1 - 7 554
          FQ230381.1 - 7 553
          FQ230619.1 - 7 553
          FQ230667.1 - 7 553
          FQ230705.1 - 7 551
          FQ230778.1 - 7 553
          FQ230864.1 - 7 553
          FQ230993.1 - 7 553
          FQ231067.1 - 7 553
          FQ231166.1 - 7 552
          FQ231444.1 - 7 552
          FQ231523.1 - 7 553
          FQ231543.1 - 7 553
          FQ231632.1 - 7 553
          FQ231815.1 - 7 535
          FQ231918.1 - 7 535
          FQ231975.1 - 7 535
          FQ232042.1 - 7 552
          FQ232191.1 - 7 553
          FQ232201.1 - 7 553
          FQ232223.1 - 7 553
          FQ232271.1 - 7 550
          FQ232364.1 - 7 555
          FQ232464.1 - 7 553
          FQ232710.1 - 7 551
          FQ232838.1 - 7 553
          FQ232847.1 - 7 553
          FQ232862.1 - 7 551
          FQ232869.1 - 7 553
          FQ232965.1 - 7 553
          FQ232977.1 - 7 553
          FQ232986.1 - 7 553
          FQ233178.1 - 7 553
          FQ233283.1 - 7 532
          FQ233669.1 - 7 552
          FQ233698.1 - 7 535
          FQ233777.1 - 7 551
          FQ233810.1 - 7 555
          FQ233845.1 - 7 553
          FQ233850.1 - 7 554
          FQ233926.1 - 7 553
          FQ234037.1 - 7 553
          FQ234069.1 - 7 553
          FQ234108.1 - 7 553
          FQ234199.1 - 7 553
          FQ234410.1 - 7 535
          FQ234418.1 - 7 553
          FQ234523.1 - 7 540
          FQ234589.1 - 7 551
          FQ234657.1 - 7 535
          FQ234685.1 - 7 553
          FQ234766.1 - 7 550
          FQ235008.1 - 7 555
          FQ235038.1 - 7 553
          FQ235049.1 - 7 535
          Y13714.1 - 8 519

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          negative regulation of endothelial cell proliferation
          signal transduction
          heart development
          response to gravity
          response to lead ion
          positive regulation of endothelial cell migration
          negative regulation of angiogenesis
          regulation of cell morphogenesis
          lung development
          response to lipopolysaccharide
          response to L-ascorbic acid
          response to cytokine
          wound healing
          regulation of cell proliferation
          response to peptide hormone
          response to ethanol
          response to cadmium ion
          inner ear development
          response to glucocorticoid
          response to cAMP
          response to calcium ion
          bone development
          cellular response to growth factor stimulus
          calcium ion binding
          collagen binding
          extracellular matrix binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0