Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPARC SIRNA Assays ›
Gene Symbol:
Sparc
Gene Name:
secreted protein, acidic, cysteine-rich (osteonectin)
Gene Aliases:
-
Chromosome Location:
Chr.10: 40742390 - 40764222 on Build Rnor_6.0
UniGene ID:
Rn.234316
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS302735
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sparc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012656.1
NP_036788.1
8
577
XM_008767663.1
XP_008765885.1
7
655
GenBank mRNA
BC061777.1
-
7
592
5621666
D28875.1
-
7
594
FQ212364.1
-
7
646
FQ212499.1
-
7
611
FQ213815.1
-
7
611
FQ213945.1
-
7
611
FQ214222.1
-
7
609
FQ214492.1
-
7
611
FQ214649.1
-
7
611
FQ214673.1
-
7
611
FQ215195.1
-
7
610
FQ215581.1
-
7
612
FQ215622.1
-
7
613
FQ215818.1
-
7
622
FQ215935.1
-
7
611
FQ216823.1
-
7
613
FQ217546.1
-
7
612
FQ219866.1
-
7
593
FQ219932.1
-
8
593
FQ219983.1
-
7
611
FQ220003.1
-
7
611
FQ220043.1
-
7
609
FQ220046.1
-
7
611
FQ220186.1
-
7
609
FQ220279.1
-
5
435
FQ220287.1
-
7
613
FQ220443.1
-
7
593
FQ220507.1
-
7
619
FQ220547.1
-
7
611
FQ220572.1
-
7
611
FQ220644.1
-
7
611
FQ220827.1
-
7
611
FQ220880.1
-
7
611
FQ220901.1
-
7
611
FQ222224.1
-
7
611
FQ223025.1
-
7
611
FQ224996.1
-
6
482
FQ225033.1
-
7
613
FQ225092.1
-
7
607
FQ225200.1
-
7
611
FQ225220.1
-
7
590
FQ225609.1
-
7
611
FQ225618.1
-
7
609
FQ225644.1
-
3
278
FQ225678.1
-
7
611
FQ225688.1
-
7
611
FQ225694.1
-
7
613
FQ225722.1
-
7
610
FQ225884.1
-
7
609
FQ225914.1
-
7
613
FQ225965.1
-
7
611
FQ226299.1
-
7
593
FQ226354.1
-
7
611
FQ226360.1
-
7
611
FQ226584.1
-
7
611
FQ226599.1
-
7
612
FQ226651.1
-
7
634
FQ226673.1
-
7
592
FQ226731.1
-
7
611
FQ226746.1
-
7
609
FQ227279.1
-
7
611
FQ227628.1
-
7
620
FQ227644.1
-
7
611
FQ227725.1
-
7
612
FQ227727.1
-
7
591
FQ227749.1
-
7
611
FQ228032.1
-
7
591
FQ228597.1
-
7
593
FQ229172.1
-
7
611
FQ229199.1
-
7
611
FQ229230.1
-
7
611
FQ229283.1
-
7
593
FQ229299.1
-
7
594
FQ229315.1
-
7
609
FQ229316.1
-
7
611
FQ230029.1
-
7
611
FQ230086.1
-
7
609
FQ230210.1
-
7
612
FQ230381.1
-
7
611
FQ230595.1
-
7
593
FQ230619.1
-
7
611
FQ230667.1
-
7
612
FQ230705.1
-
7
609
FQ230778.1
-
7
611
FQ230864.1
-
7
611
FQ230993.1
-
7
611
FQ231067.1
-
7
611
FQ231166.1
-
7
610
FQ231444.1
-
7
610
FQ231523.1
-
7
611
FQ231543.1
-
7
611
FQ231632.1
-
7
611
FQ231753.1
-
7
608
FQ231815.1
-
7
593
FQ231918.1
-
7
593
FQ231975.1
-
7
593
FQ232042.1
-
7
610
FQ232191.1
-
7
611
FQ232201.1
-
7
611
FQ232223.1
-
7
611
FQ232271.1
-
7
608
FQ232364.1
-
7
613
FQ232445.1
-
7
609
FQ232464.1
-
7
611
FQ232710.1
-
7
609
FQ232838.1
-
7
611
FQ232847.1
-
7
611
FQ232869.1
-
7
611
FQ232965.1
-
7
611
FQ232977.1
-
7
611
FQ232986.1
-
7
611
FQ233178.1
-
7
611
FQ233283.1
-
7
590
FQ233669.1
-
7
610
FQ233698.1
-
7
593
FQ233777.1
-
7
609
FQ233810.1
-
7
613
FQ233845.1
-
7
611
FQ233850.1
-
7
612
FQ233926.1
-
7
611
FQ234037.1
-
7
611
FQ234069.1
-
7
611
FQ234108.1
-
7
611
FQ234199.1
-
7
611
FQ234410.1
-
7
593
FQ234418.1
-
7
611
FQ234523.1
-
7
598
FQ234589.1
-
7
609
FQ234657.1
-
7
593
FQ234685.1
-
7
611
FQ234766.1
-
7
608
FQ235008.1
-
7
613
FQ235038.1
-
7
611
FQ235049.1
-
7
593
Y13714.1
-
8
577
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009242, AK082283, CT010306, AK148670, AK146935, and others.
Gene Ontology Categories:
Function(s)
Process(es)
ossification
negative regulation of endothelial cell proliferation
signal transduction
heart development
response to gravity
response to lead ion
positive regulation of endothelial cell migration
negative regulation of angiogenesis
regulation of cell morphogenesis
lung development
response to lipopolysaccharide
response to L-ascorbic acid
response to cytokine
wound healing
regulation of cell proliferation
response to peptide hormone
response to ethanol
response to cadmium ion
inner ear development
response to glucocorticoid
response to cAMP
response to calcium ion
bone development
cellular response to growth factor stimulus
calcium ion binding
collagen binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0