Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAP1 SIRNA Assays ›
          Gene Symbol:
          Tap1
          Gene Name:
          transporter 1, ATP-binding cassette, sub-family B (MDR/TAP)
          Gene Aliases:
          Abcb2, Cim, Tap2
          Chromosome Location:
          Chr.20: 3979302 - 3989669 on Build Rnor_6.0
          UniGene ID:
          Rn.10763
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS302753
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tap1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032055.4 NP_114444.2 1 407
          GenBank mRNA BC101854.1 - 1 674 7933924
          KC222902.1 - 1 348
          KC222903.1 - 1 348
          KC222904.1 - 1 348
          KC222905.1 - 1 165
          KC222906.1 - 1 348
          X57523.1 - 1 395
          Y10230.1 - 1 348
          Y10231.1 - 1 348
          Y10232.1 - 1 348
          Y10234.1 - 1 348
          Y10235.1 - 1 348

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          metabolic process
          protein transport
          peptide transport
          intracellular transport of viral protein in host cell
          antigen processing and presentation of endogenous peptide antigen via MHC class I
          protection from natural killer cell mediated cytotoxicity
          cytosol to ER transport
          peptide antigen transport
          transmembrane transport
          nucleotide binding
          ATP binding
          peptide transporter activity
          peptide antigen-transporting ATPase activity
          peptide-transporting ATPase activity
          ATPase activity
          MHC class Ib protein binding
          MHC class I protein binding
          peptide antigen binding
          ATPase activity, coupled to transmembrane movement of substances
          protein homodimerization activity
          ADP binding
          TAP1 binding
          TAP2 binding
          tapasin binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b776c6558-ktt9x:80/100.66.134.75:80.
          git-commit: 08b7be5c3d3493be030128aff2ceec7cf49358c5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc