Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053021.2 | NP_444180.2 | 4 | 408 | ||
XM_006252094.2 | XP_006252156.1 | 3 | 286 | |||
GenBank mRNA | BC061534.1 | - | 3 | 187 | 6886725 | |
FQ209467.1 | - | 3 | 184 | |||
FQ209498.1 | - | 3 | 185 | |||
FQ209623.1 | - | 3 | 183 | |||
FQ211071.1 | - | 3 | 183 | |||
FQ213433.1 | - | 3 | 182 | |||
FQ214098.1 | - | 3 | 228 | |||
FQ218237.1 | - | 3 | 183 | |||
FQ218812.1 | - | 3 | 183 | |||
FQ219030.1 | - | 3 | 183 | |||
M16975.1 | - | 4 | 408 | |||
M64723.1 | - | 3 | 173 | |||
X13231.1 | - | 3 | 172 |