Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012688.3 | NP_036820.1 | 3 | 707 | ||
XM_008770188.1 | XP_008768410.1 | 4 | 891 | |||
GenBank mRNA | M88096.1 | - | 3 | 711 |
Set Membership: |
![]() |