Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012750.1 | NP_036882.1 | 8 | 1360 | ||
XM_006252238.2 | XP_006252300.1 | 7 | 2129 | |||
GenBank mRNA | AF003825.1 | - | 1910 | |||
AF005226.1 | - | 8 | 1240 | |||
U97143.1 | - | 8 | 1360 |