Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017086.1 | NP_058782.1 | 2 | 1061 | ||
XM_006252239.2 | XP_006252301.1 | 2 | 1222 | |||
XM_006252240.2 | XP_006252302.1 | 3 | 1388 | |||
XM_008770818.1 | XP_008769040.1 | 2 | 1148 | |||
GenBank mRNA | U12428.1 | - | 2 | 1061 |