Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109882.1 | NP_001103352.1 | 5 | 767 | ||
NM_012783.3 | NP_036915.1 | 4 | 419 | |||
GenBank mRNA | AY120888.1 | - | 5 | 686 | ||
BC059145.1 | - | 4 | 343 | 6889785 | ||
FQ212571.1 | - | 4 | 355 | |||
FQ212582.1 | - | 4 | 363 | |||
FQ213065.1 | - | 4 | 365 | |||
FQ213626.1 | - | 4 | 355 | |||
FQ214193.1 | - | 4 | 355 | |||
FQ214232.1 | - | 4 | 360 | |||
FQ218399.1 | - | 4 | 355 | |||
FQ219910.1 | - | 4 | 339 | |||
FQ219990.1 | - | 4 | 355 | |||
FQ220920.1 | - | 4 | 363 | |||
FQ224837.1 | - | 4 | 364 | |||
FQ229755.1 | - | 4 | 352 | |||
FQ230014.1 | - | 4 | 365 | |||
FQ231850.1 | - | 4 | 365 | |||
FQ232262.1 | - | 4 | 355 | |||
FQ234420.1 | - | 4 | 365 | |||
FQ234809.1 | - | 4 | 363 | |||
X54640.1 | - | 546 | ||||
X67215.1 | - | 4 | 340 |