Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271066.1 | NP_001257995.1 | 17, 18 | 2581 | ||
NM_001271067.1 | NP_001257996.1 | 16, 17 | 2566 | |||
NM_012984.3 | NP_037116.2 | 15, 16 | 2476 | |||
XM_006252824.2 | XP_006252886.1 | 16, 17 | 2605 | |||
XM_006252825.2 | XP_006252887.1 | 16, 17 | 2605 | |||
XM_006252826.2 | XP_006252888.1 | 16, 17 | 2602 | |||
XM_006252827.2 | XP_006252889.1 | 16, 17 | 2605 | |||
XM_006252828.2 | XP_006252890.1 | 16, 17 | 2605 | |||
XM_006252829.2 | XP_006252891.1 | 16, 17 | 2605 | |||
XM_006252830.2 | XP_006252892.1 | 15, 16 | 2515 | |||
XM_006252831.2 | XP_006252893.1 | 15, 16 | 2512 | |||
XM_006252832.2 | XP_006252894.1 | 15, 16 | 2515 | |||
XM_006252833.2 | XP_006252895.1 | 15, 16 | 2512 | |||
XM_006252835.2 | XP_006252897.1 | 15, 16 | 2512 | |||
XM_006252836.2 | XP_006252898.1 | 15, 16 | 2515 | |||
XM_006252837.2 | XP_006252899.1 | 15, 16 | 2512 | |||
XM_006252839.2 | XP_006252901.1 | 16, 17 | 2605 |