Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013168.2 | NP_037300.2 | 14 | 1151 | ||
XM_006242896.1 | XP_006242958.1 | 14 | 1237 | |||
XM_006242897.1 | XP_006242959.1 | 13 | 1159 | |||
XM_008766148.1 | XP_008764370.1 | 13 | 1321 | |||
XM_008766149.1 | XP_008764371.1 | 13 | 1258 | |||
GenBank mRNA | BC088162.1 | - | 14 | 1151 | 7375228 | |
FQ225972.1 | - | 8 | 636 | |||
FQ226062.1 | - | 14 | 1074 | |||
FQ230962.1 | - | 8 | 636 | |||
FQ231091.1 | - | 14 | 1136 | |||
FQ234793.1 | - | 6 | 523 | |||
FQ234870.1 | - | 12 | 1307 | |||
X06827.1 | - | 14 | 1026 | |||
Y12006.1 | - | 14 | 1021 |