Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175577.3 | NP_783167.2 | 3 | 344 | ||
XM_006231021.2 | XP_006231083.1 | 3 | 451 | |||
XM_008760218.1 | XP_008758440.1 | 3 | 451 | |||
XM_008760219.1 | XP_008758441.1 | 3 | 450 | |||
GenBank mRNA | AF488727.1 | - | 3 | 283 | ||
AY683561.1 | - | 3 | 266 | |||
BC089767.1 | - | 3 | 344 | 7383438 |
Gene Symbol: LOC100911366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760255.1 | XP_008758477.1 | 2 | 381 |