Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017140.1 | NP_058836.1 | 6, 7 | 1213 | ||
XM_006248313.2 | XP_006248375.2 | 7, 8 | 1430 | |||
GenBank mRNA | X53944.1 | - | 6, 7 | 1213 |