Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERPINE2 SIRNA Assays ›
          Gene Symbol:
          Serpine2
          Gene Name:
          serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2
          Gene Aliases:
          Gdnpn1, PI-7, Pn-1, Spin4
          Chromosome Location:
          Chr.9: 85560852 - 85626125 on Build Rnor_6.0
          UniGene ID:
          Rn.2271
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS308643
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Serpine2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019197.1 NP_062070.1 3 573
          XM_006245161.2 XP_006245223.1 3 1236
          XM_006245162.2 XP_006245224.1 4 700
          XM_008767212.1 XP_008765434.1 3 560
          GenBank mRNA CK357674.1 - 3 573
          FQ228974.1 - 1 142
          M17784.1 - 2 449

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          embryo implantation
          blood coagulation
          negative regulation of cell proliferation
          response to radiation
          response to wounding
          negative regulation of peptidase activity
          negative regulation of platelet activation
          negative regulation of plasminogen activation
          negative regulation of sodium ion transport
          negative regulation of endopeptidase activity
          positive regulation of neuron projection development
          negative regulation of phosphatidylinositol 3-kinase signaling
          cerebellar granular layer morphogenesis
          negative regulation of blood coagulation
          negative regulation of cell growth
          secretion by cell
          secretory granule organization
          negative regulation of protein catabolic process
          mating plug formation
          negative regulation of proteolysis
          negative regulation of smoothened signaling pathway
          regulation of timing of cell differentiation
          positive regulation of astrocyte differentiation
          detection of mechanical stimulus involved in sensory perception
          regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          innervation
          seminal vesicle epithelium development
          dense core granule biogenesis
          negative regulation of platelet aggregation
          serine-type endopeptidase inhibitor activity
          receptor binding
          glycosaminoglycan binding
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0