Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019621.1 | NP_062567.1 | 1, 2 | 76 | ||
XM_006246686.2 | XP_006246748.1 | 1, 2 | 132 | |||
XM_006246687.2 | XP_006246749.1 | 1, 2 | 131 | |||
GenBank mRNA | M96853.1 | - | 1, 2 | 76 | ||
X66474.1 | - | 1, 2 | 221 |