Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2B4 SIRNA Assays ›
          Gene Symbol:
          Atp2b4
          Gene Name:
          ATPase, Ca++ transporting, plasma membrane 4
          Gene Aliases:
          PMCA4
          Chromosome Location:
          Chr.13: 50091639 - 50153808 on Build Rnor_6.0
          UniGene ID:
          Rn.9986
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS310342
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp2b4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005871.1 NP_001005871.1 1 314
          XM_008769446.1 XP_008767668.1 2 697
          XM_008769447.1 XP_008767669.1 2 1334
          XM_008769448.1 XP_008767670.1 2 697
          XM_008769449.1 XP_008767671.1 2 698
          XM_008769450.1 XP_008767672.1 2 698
          GenBank mRNA U15408.1 - 1 314

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neural retina development
          regulation of transcription from RNA polymerase II promoter
          calcium ion transport
          cellular calcium ion homeostasis
          spermatogenesis
          metabolic process
          negative regulation of nitric oxide mediated signal transduction
          hippocampus development
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of nitric oxide biosynthetic process
          negative regulation of nitric-oxide synthase activity
          response to hydrostatic pressure
          calcium ion transmembrane transport
          negative regulation of calcineurin-NFAT signaling cascade
          cellular response to epinephrine stimulus
          negative regulation of the force of heart contraction
          negative regulation of arginine catabolic process
          negative regulation of adrenergic receptor signaling pathway involved in heart process
          calcium ion export
          negative regulation of cardiac muscle hypertrophy in response to stress
          negative regulation of citrulline biosynthetic process
          positive regulation of cAMP-dependent protein kinase activity
          calcium-transporting ATPase activity
          calmodulin binding
          ATP binding
          sodium channel regulator activity
          PDZ domain binding
          protein phosphatase 2B binding
          nitric-oxide synthase inhibitor activity
          metal ion binding
          nitric-oxide synthase binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix