Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037766.2 | NP_001032855.2 | 40 | 5820 | ||
GenBank mRNA | FQ227992.1 | - | 1 | 85 | ||
FQ230570.1 | - | 1 | 85 | |||
FQ235231.1 | - | 1 | 85 |