Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013924.1 | NP_001013946.1 | 8 | 1168 | ||
XM_006235817.2 | XP_006235879.1 | 8 | 1594 | |||
XM_006235818.2 | XP_006235880.1 | 8 | 1214 | |||
XM_006235820.2 | XP_006235882.1 | 8 | 1222 | |||
XM_008762485.1 | XP_008760707.1 | 7 | 2728 | |||
XM_008762486.1 | XP_008760708.1 | 8 | 1598 | |||
XM_008762487.1 | XP_008760709.1 | 9 | 1315 | |||
XM_008762488.1 | XP_008760710.1 | 9 | 1338 | |||
XM_008762489.1 | XP_008760711.1 | 8 | 1237 | |||
XM_008762490.1 | XP_008760712.1 | 9 | 1294 | |||
XM_008762491.1 | XP_008760713.1 | 8 | 1218 | |||
XM_008762492.1 | XP_008760714.1 | 6 | 2509 | |||
XM_008762493.1 | XP_008760715.1 | 8 | 1245 | |||
XM_008762494.1 | XP_008760716.1 | 7 | 2721 | |||
XM_008762495.1 | XP_008760717.1 | 6 | 2589 | |||
GenBank mRNA | BC083707.1 | - | 8 | 1168 | 7190542 |