Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006978.1 | NP_001006979.1 | 3 | 130 | ||
GenBank mRNA | BC084705.1 | - | 3 | 130 | 7301091 | |
FQ212944.1 | - | 3 | 150 | |||
FQ216715.1 | - | 3 | 148 |