Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam96a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008327.1 | NP_001008328.1 | 1 | 233 | ||
GenBank mRNA | BC086524.1 | - | 1 | 233 | 7303322 | |
FQ210328.1 | - | 1 | 214 | |||
FQ215407.1 | - | 1 | 270 | |||
FQ216067.1 | - | 1 | 277 | |||
FQ216458.1 | - | 1 | 271 | |||
FQ216583.1 | - | 1 | 231 | |||
FQ218392.1 | - | 1 | 214 | |||
FQ218618.1 | - | 1 | 277 | |||
FQ220244.1 | - | 1 | 193 | |||
FQ225271.1 | - | 1 | 130 | |||
FQ232448.1 | - | 1 | 231 | |||
FQ233915.1 | - | 1 | 138 |