Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035007.1 | NP_001030179.1 | 11 | 1638 | ||
XM_006256944.2 | XP_006257006.1 | 9 | 1327 |