Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107047.1 | NP_001100517.1 | 8 | 1367 | ||
XM_008768049.1 | XP_008766271.1 | 3 | 1464 | |||
XM_008768050.1 | XP_008766272.1 | 3 | 1338 |