Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL6 SIRNA Assays ›
          Gene Symbol:
          Bcl6
          Gene Name:
          B-cell CLL/lymphoma 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 80255722 - 80279077 on Build Rnor_6.0
          UniGene ID:
          Rn.20571
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS314648
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bcl6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001107084.1 NP_001100554.1 6 1711
          XM_008768799.1 XP_008767021.1 5 2584
          GenBank mRNA BC166425.1 - 6 1611 7097735

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          negative regulation of cell-matrix adhesion
          germinal center formation
          negative regulation of type 2 immune response
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          Rho protein signal transduction
          spermatogenesis
          protein localization
          biological_process
          negative regulation of cell proliferation
          actin cytoskeleton organization
          B cell differentiation
          negative regulation of cell growth
          positive regulation of B cell proliferation
          positive regulation of histone deacetylation
          negative regulation of mast cell cytokine production
          negative regulation of Rho protein signal transduction
          type 2 immune response
          regulation of cell proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of memory T cell differentiation
          negative regulation of cell differentiation
          negative regulation of T-helper 2 cell differentiation
          positive regulation of neuron differentiation
          negative regulation of Notch signaling pathway
          negative regulation of transcription, DNA-templated
          negative regulation of isotype switching to IgE isotypes
          erythrocyte development
          regulation of inflammatory response
          positive regulation of cellular component movement
          negative regulation of cellular senescence
          RNA polymerase II regulatory region sequence-specific DNA binding
          intronic transcription regulatory region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          molecular_function
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          chromatin DNA binding
          sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix