Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047093.1 | NP_001040558.1 | 1 | 443 | ||
GenBank mRNA | AY641809.1 | - | 1 | 443 |