Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFT88 SIRNA Assays ›
          Gene Symbol:
          Ift88
          Gene Name:
          intraflagellar transport 88
          Gene Aliases:
          Ttc10
          Chromosome Location:
          Chr.15: 37690412 - 37785877 on Build Rnor_6.0
          UniGene ID:
          Rn.132110
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS315927
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ift88
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001107266.1 NP_001100736.1 10, 11 768
          XM_006252066.2 XP_006252128.1 10, 11 1750
          XM_006252067.2 XP_006252129.1 11, 12 1942
          XM_006252068.2 XP_006252130.1 10, 11 1051
          XM_006252070.2 XP_006252132.1 10, 11 1749
          XM_006252071.2 XP_006252133.1 10, 11 1750

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009376, L31959, BC012250, AK168395, AK135110.

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          kidney development
          liver development
          epithelial cell morphogenesis
          Notch signaling pathway
          smoothened signaling pathway
          sperm axoneme assembly
          spermatid nucleus elongation
          determination of left/right symmetry
          nervous system development
          brain development
          heart development
          protein localization
          epidermis development
          regulation of smoothened signaling pathway
          organ morphogenesis
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          spinal cord dorsal/ventral patterning
          telencephalon development
          lung development
          pancreas development
          cytoplasmic microtubule organization
          response to fluid shear stress
          epidermal stem cell homeostasis
          cilium assembly
          regulation of odontogenesis of dentin-containing tooth
          embryonic digit morphogenesis
          positive regulation of insulin-like growth factor receptor signaling pathway
          regulation of fat cell differentiation
          forebrain morphogenesis
          negative regulation of epithelial cell proliferation
          cardiac muscle cell differentiation
          palate development
          inner ear receptor stereocilium organization
          limb development
          regulation of feeding behavior
          cilium morphogenesis
          cardiac septum morphogenesis
          lung vasculature development
          heart formation
          regulation of protein processing
          cochlea development
          regulation of cilium assembly
          regulation of autophagosome assembly
          kinesin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix