Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182821.1 | NP_877973.1 | 4 | 414 | ||
XM_006251775.1 | XP_006251837.1 | 4 | 442 | |||
XM_006251776.2 | XP_006251838.1 | 4 | 442 | |||
XM_008770531.1 | XP_008768753.1 | 2 | 932 | |||
XM_008770532.1 | XP_008768754.1 | 2 | 932 | |||
XM_008770533.1 | XP_008768755.1 | 2 | 932 | |||
XM_008770534.1 | XP_008768756.1 | 2 | 932 | |||
XM_008770535.1 | XP_008768757.1 | 2 | 932 | |||
XM_008770536.1 | XP_008768758.1 | 2 | 932 | |||
XM_008770537.1 | XP_008768759.1 | 2 | 932 | |||
XM_008770538.1 | XP_008768760.1 | 2 | 932 | |||
XM_008770540.1 | XP_008768762.1 | 3 | 307 | |||
GenBank mRNA | AY327408.1 | - | 4 | 414 | ||
BC098901.1 | - | 4 | 356 | 7127537 |