Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034104.1 | NP_001029276.1 | 878 | |||
GenBank mRNA | BC100657.1 | - | 878 | 7936370 |
Gene Symbol: LOC103690093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758838.1 | XP_008757060.1 | 6 | 1062 |