Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212520.1 | NP_997685.1 | 6 | 744 | ||
XM_006228420.2 | XP_006228482.1 | 7 | 852 | |||
GenBank mRNA | AB162855.1 | - | 6 | 744 | ||
FQ226953.1 | - | 3 | 262 |