Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam189b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107690.1 | NP_001101160.1 | 12 | 2370 | ||
XM_006232656.1 | XP_006232718.1 | 12 | 2833 | |||
XM_006232664.1 | XP_006232726.1 | 12 | 2779 | |||
XM_008761161.1 | XP_008759383.1 | 11 | 2968 | |||
XM_008761162.1 | XP_008759384.1 | 11 | 2965 | |||
XM_008761163.1 | XP_008759385.1 | 11 | 2956 | |||
XM_008761164.1 | XP_008759386.1 | 11 | 2935 | |||
XM_008761165.1 | XP_008759387.1 | 11 | 2926 | |||
XM_008761166.1 | XP_008759388.1 | 11 | 2923 | |||
XM_008761167.1 | XP_008759389.1 | 12 | 2821 | |||
XM_008761168.1 | XP_008759390.1 | 12 | 2818 | |||
XM_008761169.1 | XP_008759391.1 | 12 | 2791 | |||
GenBank mRNA | FQ221099.1 | - | 12 | 2373 |