Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239085.2 | XP_006239147.1 | 12 | 1998 | ||
XM_006239086.2 | XP_006239148.1 | 12 | 1998 | |||
XM_006239087.2 | XP_006239149.1 | 12 | 3950 | |||
XM_006239088.2 | XP_006239150.1 | 12 | 1998 | |||
XM_006239089.2 | XP_006239151.1 | 12 | 1998 | |||
XM_006239090.2 | XP_006239152.1 | 12 | 1998 | |||
XM_006239091.2 | XP_006239153.1 | 12 | 1998 | |||
XM_006239092.2 | XP_006239154.1 | 12 | 1999 | |||
XM_006239093.2 | XP_006239155.1 | 12 | 1834 | |||
XM_006239094.2 | XP_006239156.1 | 12 | 1998 | |||
XM_006239095.1 | XP_006239157.1 | 12 | 1857 | |||
XM_006239096.2 | XP_006239158.1 | 12 | 1996 | |||
XM_006239097.2 | XP_006239159.1 | 12 | 1996 | |||
XM_006239098.2 | XP_006239160.1 | 12 | 1996 | |||
XM_006239099.2 | XP_006239161.1 | 12 | 1996 | |||
XM_006239100.2 | XP_006239162.1 | 12 | 1995 | |||
XM_006239101.2 | XP_006239163.1 | 12 | 1997 | |||
XM_006239103.2 | XP_006239165.1 | 12 | 1995 | |||
XM_006239104.2 | XP_006239166.1 | 12 | 1996 | |||
XM_006239105.2 | XP_006239167.1 | 12 | 1998 | |||
XM_008764184.1 | XP_008762406.1 | 12 | 1995 | |||
XM_008764185.1 | XP_008762407.1 | 12 | 1992 | |||
XM_232771.8 | XP_232771.6 | 12 | 1834 |