Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108239.1 | NP_001101709.1 | 4 | 494 | ||
XM_008767114.1 | XP_008765336.1 | 3 | 523 | |||
XM_008767115.1 | XP_008765337.1 | 4 | 557 |