Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108246.1 | NP_001101716.1 | 7 | 662 | ||
XM_006256652.2 | XP_006256714.1 | 7 | 698 | |||
XM_006256653.2 | XP_006256715.1 | 7 | 695 | |||
GenBank mRNA | BC085914.1 | - | 6 | 743 | ||
FQ211849.1 | - | 7 | 677 |