Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271026.1 | NP_001257955.1 | 2 | 85 | ||
GenBank mRNA | AY387053.1 | - | 531 | |||
BC089894.1 | - | 2 | 85 | |||
BC158679.1 | - | 2 | 85 | |||
FQ227271.1 | - | 2 | 106 |