Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007739.1 | NP_001007740.1 | 2 | 153 | ||
XM_006256450.2 | XP_006256512.1 | 3 | 147 | |||
XM_006256451.2 | XP_006256513.1 | 2 | 171 | |||
GenBank mRNA | BC079228.1 | - | 2 | 153 | 7132455 | |
FQ232554.1 | - | 2 | 107 |