Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013198.2 | NP_001013216.2 | 13 | 1967 | ||
GenBank mRNA | BC091427.1 | - | 7 | 853 | 7366491 |