Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX2 SIRNA Assays ›
Gene Symbol:
Rbfox2
Gene Name:
RNA binding protein, fox-1 homolog (C. elegans) 2
Gene Aliases:
Rbm9
Chromosome Location:
Chr.7: 118157093 - 118396729 on Build Rnor_6.0
UniGene ID:
Rn.137551
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS325700
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rbfox2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079895.1
NP_001073364.1
3
910
XM_006241828.2
XP_006241890.1
4
962
XM_006241829.2
XP_006241891.1
4
962
XM_006241830.2
XP_006241892.1
4
959
XM_006241831.2
XP_006241893.1
4
962
XM_006241832.2
XP_006241894.1
4
959
XM_006241833.2
XP_006241895.1
4
962
XM_006241834.2
XP_006241896.1
4
962
XM_006241835.2
XP_006241897.1
4
962
XM_006241836.2
XP_006241898.1
4
962
XM_006241837.2
XP_006241899.1
4
959
XM_006241838.2
XP_006241900.1
4
962
XM_006241839.2
XP_006241901.1
4
962
XM_006241840.2
XP_006241902.1
4
962
XM_006241841.2
XP_006241903.1
4
962
XM_006241842.2
XP_006241904.1
4
3475
XM_006241843.2
XP_006241905.1
4
959
XM_006241845.2
XP_006241907.1
3
547
XM_006241846.2
XP_006241908.1
3
547
XM_006241847.2
XP_006241909.1
3
546
XM_006241848.2
XP_006241910.1
3
438
XM_006241849.2
XP_006241911.1
4
962
XM_006241850.2
XP_006241912.1
4
962
XM_008765584.1
XP_008763806.1
4
985
XM_008765585.1
XP_008763807.1
4
962
XM_008765586.1
XP_008763808.1
4
962
GenBank mRNA
BC128766.1
-
3
910
7123798
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_053104, NM_001110828, NM_001286419, NM_001110830, NM_001110827, and others.
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
RNA splicing
regulation of definitive erythrocyte differentiation
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix