Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047916.1 | NP_001041381.1 | 8, 9 | 1039 | ||
XM_006243513.2 | XP_006243575.1 | 1777 | ||||
XM_006243514.1 | XP_006243576.1 | 1594 | ||||
XM_006243515.1 | XP_006243577.1 | 1732 | ||||
XM_006243516.1 | XP_006243578.1 | 1732 | ||||
XM_006243517.2 | XP_006243579.1 | 7, 8 | 1004 | |||
XM_006243518.2 | XP_006243580.1 | 9, 10 | 1228 | |||
XM_006243519.2 | XP_006243581.1 | 9, 10 | 1228 | |||
GenBank mRNA | AY325202.1 | - | 8, 9 | 1039 |
Gene Symbol: LOC100910069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766471.1 | XP_008764693.1 | 11, 12 | 1819 |