Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017327.1 | NP_059023.1 | 7 | 1444 | ||
XM_008772320.1 | XP_008770542.1 | 7 | 1453 | |||
XM_008772322.1 | XP_008770544.1 | 6 | 4062 | |||
GenBank mRNA | AF413211.1 | - | 2 | 42 | ||
AF413212.1 | - | 2 | 42 | |||
M12671.1 | - | 6 | 608 | |||
M17526.1 | - | 7 | 1444 |