Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PITX2 SIRNA Assays ›
          Gene Symbol:
          Pitx2
          Gene Name:
          paired-like homeodomain 2
          Gene Aliases:
          Pitx-2, Pitx2c, Ptx2
          Chromosome Location:
          Chr.2: 233602732 - 233621055 on Build Rnor_6.0
          UniGene ID:
          Rn.17591
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS329041
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pitx2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042505.1 NP_001035970.1 3 900
          NM_019334.2 NP_062207.1 4 601
          GenBank mRNA AF039832.1 - 4 601
          AJ222971.1 - 3 908
          AJ222972.1 - 4 654
          EF519321.1 - 3 390

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          patterning of blood vessels
          vasculogenesis
          in utero embryonic development
          neuron migration
          extraocular skeletal muscle development
          atrioventricular valve development
          cardiac neural crest cell migration involved in outflow tract morphogenesis
          pulmonary myocardium development
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          determination of left/right symmetry
          brain development
          heart development
          skeletal muscle tissue development
          myoblast fusion
          male gonad development
          female gonad development
          anatomical structure morphogenesis
          response to hormone
          organ morphogenesis
          Wnt signaling pathway
          subthalamic nucleus development
          hypothalamus cell migration
          pituitary gland development
          neuron differentiation
          lung development
          regulation of cell migration
          embryonic camera-type eye development
          response to vitamin A
          embryonic hindlimb morphogenesis
          vascular smooth muscle cell differentiation
          deltoid tuberosity development
          regulation of cell proliferation
          odontogenesis of dentin-containing tooth
          odontogenesis
          camera-type eye development
          positive regulation of DNA binding
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          spleen development
          embryonic digestive tract morphogenesis
          cardiac muscle tissue development
          cardiac muscle cell differentiation
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle cell development
          digestive system development
          ventricular septum morphogenesis
          left lung morphogenesis
          pulmonary vein morphogenesis
          superior vena cava morphogenesis
          endodermal digestive tract morphogenesis
          iris morphogenesis
          cell proliferation involved in outflow tract morphogenesis
          left/right axis specification
          positive regulation of myoblast proliferation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          transcriptional repressor activity, RNA polymerase II transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor binding
          chromatin DNA binding
          identical protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix