Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020082.2 | NP_064467.1 | 2 | 495 | ||
XM_006251906.2 | XP_006251968.1 | 2 | 534 | |||
XM_006251907.2 | XP_006251969.1 | 2 | 4112 | |||
GenBank mRNA | AF041066.1 | - | 2 | 506 | ||
BC070953.1 | - | 2 | 495 | 7103267 | ||
BC166436.1 | - | 2 | 1078 | 7372999 |