Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020093.1 | NP_064478.1 | 1178 | |||
GenBank mRNA | AB039878.1 | - | 1178 | |||
AF168004.1 | - | 1198 | ||||
AF210434.1 | - | 1170 | ||||
AF257234.1 | - | 1108 | ||||
AF343574.1 | - | 867 | ||||
AF343575.1 | - | 732 | ||||
AF381277.1 | - | 1180 | ||||
AF381278.1 | - | 1108 | ||||
AF381279.1 | - | 986 | ||||
AF381280.1 | - | 1264 | ||||
AF381281.1 | - | 1024 | ||||
AF381285.1 | - | 244 | ||||
GU322017.1 | - | 1277 | ||||
GU322019.1 | - | 1208 | ||||
GU322363.1 | - | 1037 | ||||
GU322364.1 | - | 1121 | ||||
GU345835.1 | - | 1355 | ||||
GU345836.1 | - | 1156 | ||||
KC774169.1 | - | 964 | ||||
KC774170.1 | - | 964 | ||||
KC774172.1 | - | 1118 | ||||
KC774173.1 | - | 752 | ||||
KC774174.1 | - | 401 | ||||
KC774175.1 | - | 818 | ||||
KC774176.1 | - | 765 | ||||
KC774177.1 | - | 546 |