Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020093.1 | NP_064478.1 | 2 | 151 | ||
GenBank mRNA | AB039878.1 | - | 2 | 151 | ||
AF210434.1 | - | 2 | 143 | |||
AF257234.1 | - | 1 | 81 | |||
AF343574.1 | - | 1 | 81 | |||
AF381277.1 | - | 1 | 81 | |||
AF381278.1 | - | 1 | 81 | |||
AF381279.1 | - | 1 | 81 | |||
AF381280.1 | - | 1 | 81 | |||
AF381281.1 | - | 1 | 81 | |||
GU322017.1 | - | 2 | 94 | |||
GU322019.1 | - | 2 | 94 | |||
GU322363.1 | - | 2 | 94 | |||
GU322364.1 | - | 2 | 94 | |||
GU345835.1 | - | 2 | 94 | |||
GU345836.1 | - | 2 | 94 | |||
KC774176.1 | - | 1 | 101 |