Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021694.3 | NP_067726.3 | 14, 15 | 1532 | ||
GenBank mRNA | AF314539.1 | - | 12, 13 | 1443 | ||
AJ236911.1 | - | 12, 13 | 1423 | |||
BC091218.1 | - | 14, 15 | 1532 | 7107802 |