Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
Pcn, Plec1, Pltn
Chromosome Location:
Chr.7: 117230319 - 117291473 on Build Rnor_6.0
UniGene ID:
Rn.1085
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS329852
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164296.1
NP_001157768.1
14
1765
NM_001164297.1
NP_001157769.1
14
1700
NM_001164298.1
NP_001157770.1
14
1759
NM_001164299.1
NP_001157771.1
14
1791
NM_001164302.1
NP_001157774.1
14
1745
NM_001164303.1
NP_001157775.1
14
1698
NM_001164304.1
NP_001157776.1
14
1958
NM_001164305.1
NP_001157777.1
14
1675
NM_001164307.1
NP_001157779.1
14
1772
NM_001164308.1
NP_001157780.1
14
1832
NM_022401.2
NP_071796.2
14
2151
XM_006241870.2
XP_006241932.1
15
2362
XM_006241875.2
XP_006241937.1
15
2124
XM_006241882.2
XP_006241944.1
15
2479
XM_008765592.1
XP_008763814.1
15
2955
XM_008765593.1
XP_008763815.1
16
2139
XM_008765594.1
XP_008763816.1
15
3122
XM_008765595.1
XP_008763817.1
15
3926
XM_008765596.1
XP_008763818.1
15
1749
XM_008765597.1
XP_008763819.1
15
1872
XM_008765598.1
XP_008763820.1
15
2362
GenBank mRNA
AY480022.1
-
14
1768
AY480023.1
-
14
1703
AY480024.1
-
14
1762
AY480025.1
-
14
1794
AY480026.1
-
14
1748
AY480027.1
-
14
2295
AY480028.1
-
14
1701
AY480029.1
-
14
1846
AY480030.1
-
14
1678
AY480031.1
-
14
1775
AY480032.1
-
14
1835
X59601.1
-
14
2101
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
female pregnancy
response to nutrient
epithelial cell differentiation
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline