Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022860.2 | NP_074051.1 | 11 | 1566 | ||
XM_006241497.2 | XP_006241559.1 | 11 | 2093 | |||
XM_008765419.1 | XP_008763641.1 | 11 | 3922 | |||
XM_008765420.1 | XP_008763642.1 | 10 | 2243 | |||
GenBank mRNA | BC081799.1 | - | 11 | 1566 | 7133837 | |
D17809.1 | - | 11 | 1455 |
Gene Symbol: Slc26a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241491.2 | XP_006241553.1 | 18 | 4614 | ||
GenBank mRNA | FQ211772.1 | - | 18 | 2904 |